Learn More
OHAUS™ Defender™ 5000 R Series Base
Product Code. p-7162284 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
15.00 kg, 30.00 kg
150.00 kg, 300.00 kg
3.00 kg, 6.00 kg
30.00 kg, 60.00 kg
300.00 kg, 600.00 kg
6.00 kg, 15.00 kg
60.00 kg, 150.00 kg
Model:
D150RQDV
D150RQDX
D150RTDV
D15RQDL
D15RQDR
D300RQDV
D300RTDV
D30RQDL
D600RTDV
D60RQDL
D60RQDV
D60RQDX
D6RQDR
Unit Size:
1 unit
Product Code. Capacity Model unitSize
15908435 300.00 kg, 600.00 kg D600RTDV 1 unit
15928415 6.00 kg, 15.00 kg D15RQDL 1 unit
15928425 30.00 kg, 60.00 kg D60RQDV 1 unit
15938415 15.00 kg, 30.00 kg D30RQDL 1 unit
15938425 60.00 kg, 150.00 kg D150RQDV 1 unit
15948415 30.00 kg, 60.00 kg D60RQDL 1 unit
15948425 150.00 kg, 300.00 kg D300RQDV 1 unit
15988405 3.00 kg, 6.00 kg D6RQDR 1 unit
15988415 30.00 kg, 60.00 kg D60RQDX 1 unit
15988425 60.00 kg, 150.00 kg D150RTDV 1 unit
15998405 6.00 kg, 15.00 kg D15RQDR 1 unit
15998415 60.00 kg, 150.00 kg D150RQDX 1 unit
15998425 150.00 kg, 300.00 kg D300RTDV 1 unit
13 options
Product Code. 15908435
Brand: OHAUS™ 30419624
OHAUS™ Defender™ 5000 R Series Base offers durability for demanding industrial applications. The R series features moisture-protected single-point load cells, powder-coated steel construction, non-slip rubber adjustable feet, removable stainless steel weighing pans and leveling bubbles.
  • The R Series features a stainless steel platform, powder-coated steel frame and an aluminum load cell to withstand harsh industrial environments.
  • The Defender 5000 bases are equipped with an EasyConnect cable, which allows instant configuration with Defender 5000 (TD52) indicators, speeding up installation.
  • Defender 5000 bases are NTEP-certified and Measurement Canada approved.
  • These bases also carry OIML and EC Type approvals, which make them ideal for legal-for-trade use.
TRUSTED_SUSTAINABILITY
Specifications
Construction 304-grade stainless steel platform, sand grey powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet, level indicator
Capacity 300.00 kg, 600.00 kg
Readability 20.000000 g, 50.000000 g
Model D600RTDV
Protection Class IP67
Temperature (Metric) Operating -10°C to +40°C
Dimensions (L x W x H) 821 x 600 x 132 mm
Weight (Metric) 39.000 kg