Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale (EU), Certified
Bench Scales for Basic Industrial Applications
8290.00 NOK - 13645.00 NOK
Specifications
Battery Life | 100 Hours with Rechargeable Battery |
---|---|
Battery Type | AC Adapter (Included);Rechargeable Battery (Included) |
Connection Type | Standard RS232 and optional USB Device or Ethernet |
Construction | Stainless steel indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet |
Display Type | 7 Segment LCD with Multicolor Backlight |
Product Code | Brand | Model | Readability | Capacity | Dimensions (L x W x H) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Readability | Capacity | Dimensions (L x W x H) | Price | Quantity & Availability | ||
17525845
|
OHAUS™
30685262 |
D33XW15B1R1EU | 5 g | 15 kg | 489 x 305 x 569 mm |
8290.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17535845
|
OHAUS™
30685263 |
D33XW30B1R1EU | 10 g | 30 kg | 489 x 305 x 569 mm |
8520.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17545845
|
OHAUS™
30685264 |
D33XW60B1R1EU | 20 g | 60 kg | 489 x 305 x 569 mm |
8520.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17555845
|
OHAUS™
30685265 |
D33XW60B1L2EU | 20 g | 60 kg | 686 x 420 x 922 mm |
9215.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17565845
|
OHAUS™
30685266 |
D33XW150B1L2EU | 50 g | 150 kg | 686 x 420 x 922 mm |
9215.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17575845
|
OHAUS™
30685267 |
D33XW150B1X2EU | 50 g | 150 kg | 787 x 500 x 922 mm |
10710.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17585845
|
OHAUS™
30685268 |
D33XW300B1X2EU | 100 g | 300 kg | 787 x 500 x 922 mm |
10710.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17595845
|
OHAUS™
30685269 |
D33XW300B1V3EU | 100 g | 300 kg | 935 x 600 x 1222 mm |
13645.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17505855
|
OHAUS™
30685270 |
D33XW600B1V3EU | 200 g | 600 kg | 935 x 600 x 1222 mm |
13645.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17515855
|
OHAUS™
30685271 |
D33XW15B1R5EU | 5 g | 15 kg | 646 x 305 x 122 mm |
8520.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17525855
|
OHAUS™
30685272 |
D33XW30B1R5EU | 10 g | 30 kg | 646 x 305 x 122 mm |
8520.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17535855
|
OHAUS™
30685273 |
D33XW60B1R5EU | 20 g | 60 kg | 646 x 305 x 122 mm |
8520.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17545855
|
OHAUS™
30685274 |
D33XW60B1L5EU | 20 g | 60 kg | 842 x 420 x 134 mm |
9790.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17555855
|
OHAUS™
30685275 |
D33XW150B1L5EU | 50 g | 150 kg | 842 x 420 x 134 mm |
9790.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17565855
|
OHAUS™
30685276 |
D33XW150B1X5EU | 50 g | 150 kg | 941 x 500 x 150 mm |
11230.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
17575855
|
OHAUS™
30685277 |
D33XW300B1X5EU | 100 g | 300 kg | 941 x 500 x 150 mm |
11230.00 NOK
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Results are clearly shown using the large, bright backlit LCD display.
- In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
- This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
- Indicator features a stainless steel housing and mounting bracket.
- Features software for Parts Counting, Checkweighing and Totalization.
- Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Specifications
100 Hours with Rechargeable Battery | |
Standard RS232 and optional USB Device or Ethernet | |
7 Segment LCD with Multicolor Backlight | |
Weighing, Checkweighing, Counting, Totalization. |
AC Adapter (Included);Rechargeable Battery (Included) | |
Stainless steel indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet | |
-10°C to +40°C, 85%RH, non-condensing |