Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale
Product Code. 17572126 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
15 kg
30 kg
60 kg
150 kg
300 kg
600 kg
Model:
D33P150B1L2
D33P150B1L5
D33P150B1X2
D33P150B1X5
D33P15B1R1
D33P15B1R5
D33P300B1V3
D33P300B1X2
D33P300B1X5
D33P30B1R1
D33P30B1R5
D33P600B1V3
D33P60B1L2
D33P60B1L5
D33P60B1R1
D33P60B1R5
Readability:
2 g
5 g
10 g
20 g
50 g
100 g
Dimensions (L x W x H):
489 x 305 x 542 mm
646 x 305 x 130 mm
686 x 420 x 896 mm
785 x 500 x 896 mm
842 x 420 x 134 mm
941 x 500 x 150 mm
933 x 600 x 1195 mm
Unit Size:
Each
Product Code. Capacity Model Readability Dimensions (L x W x H) unitSize
17572126 300 kg D33P300B1X2 50 g 785 x 500 x 896 mm Each
17512126 15 kg D33P15B1R1 2 g 489 x 305 x 542 mm Each
17522126 30 kg D33P30B1R1 5 g 489 x 305 x 542 mm Each
17532126 60 kg D33P60B1R1 10 g 489 x 305 x 542 mm Each
17542126 60 kg D33P60B1L2 10 g 686 x 420 x 896 mm Each
17552126 150 kg D33P150B1L2 20 g 686 x 420 x 896 mm Each
17562126 150 kg D33P150B1X2 20 g 785 x 500 x 896 mm Each
17582126 300 kg D33P300B1V3 50 g 933 x 600 x 1195 mm Each
17592126 600 kg D33P600B1V3 100 g 933 x 600 x 1195 mm Each
17502136 15 kg D33P15B1R5 2 g 646 x 305 x 130 mm Each
17512136 30 kg D33P30B1R5 5 g 646 x 305 x 130 mm Each
17522136 60 kg D33P60B1R5 10 g 646 x 305 x 130 mm Each
17532136 60 kg D33P60B1L5 10 g 842 x 420 x 134 mm Each
17542136 150 kg D33P150B1L5 20 g 842 x 420 x 134 mm Each
17552136 150 kg D33P150B1X5 20 g 941 x 500 x 150 mm Each
17562136 300 kg D33P300B1X5 50 g 941 x 500 x 150 mm Each
16 options
Product Code. 17572126
Brand: OHAUS™ 30684749
Bench Scales for Basic Industrial Applications
  • Results are clearly shown using the large, bright backlit LCD display.
  • In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
  • This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
  • Indicator features a stainless steel housing and mounting bracket.
  • Features software for Parts Counting, Checkweighing and Totalization.
  • Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 100 Hours with Rechargeable Battery;360 Hours with Disposable Batteries
Battery Type 6 C (LR14) Batteries (Not Included);AC Adapter (Included);Rechargeable Battery (Not Included)
Connection Type Standard RS232 and optional USB Device or Ethernet
Construction ABS plastic indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet
Capacity 300 kg
Display Type 7-segment Multi-color Backlit LCD
Dimensions (LxW) Platform 650 x 500 mm
Readability 50 g
Model D33P300B1X2
Temperature (Metric) Operating -10°C to +40°C, 85%RH, non-condensing
Weighing Modes Weighing, Checkweighing, Counting, Totalization.
Dimensions (L x W x H) 785 x 500 x 896 mm
Weight (Metric) 27.6 kg