Learn More
Ohaus™ Defender™ 2000-D24P Bench Scale
Shop All Ohaus Products
Click to view available options
Capacity:
150 g
300 g
Unit Size:
Each
Description
- Sturdy industrial design that can stand up to the rigors of industrial weighing
- Easy-to-read red LED display can be viewed in dim conditions
- Versatile power capabilities provide unrestricted operation
Specifications
Specifications
| Battery Life | 80 Hours with Rechargeable Battery |
| Battery Type | Rechargeable |
| Connection Type | RS232 (Available as an Accessory) |
| Construction | ABS indicator, 430 grade stainless steel platform with painted steel tubular frame, IP65 aluminum load cell |
| Capacity | 150 g |
| Display Type | Red LED |
| Readability | 20.000000 g |
| Includes | AC Adapter (Included); Rechargeable Battery (Included) |
| Material | ABS indicator, 430 grade stainless steel platform with painted steel tubular frame, IP65 aluminum load cell |
| Model | D24PE150FX |
| Show More |