Learn More
Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators
Product Code. 10214832 Shop All Thermo Scientific Products
Click to view available options
Displacement:
116L/min. (at 60Hz); 97L/min. (at 50Hz)
195L/min. (at 60Hz); 162L/min. (at 50Hz)
283L/min. (at 60Hz); 237L/min. (at 50Hz)
76L/min. (at 60Hz); 62L/min. (at 50Hz)
Voltage:
230 V
Unit Size:
Each
Product Code. 10214832
Brand: Thermo Scientific™ VLP200230
Only null left
Add to basket
Add to basket
Achieve maximum solute recovery with the Thermo Scientific™Deep Vacuum Oil Pump.
Achieve maximum solute recovery with the Thermo Scientific™ Deep Vacuum Oil Pumps for Vacuum Concentrators, designed to work with all Thermo Scientific™ SpeedVac™ concentrating and freeze drying systems.
Features:
  • Three-position gas ballast control optimizes pump performance to suit all applications
  • Large, sideview sight glass allows for convenient checking of proper oil levels at a glance
All Rotary Vane Oil Pumps come standard with:
  • Muffler (OME190 Oil Mist Eliminator)
  • Two Oil Filters (MF190)
  • Quick Drain Kit
TRUSTED_SUSTAINABILITY
Specifications
Description VLP200; 230V 50Hz
Type Deep Vacuum Oil Pump
Displacement 195L/min. (at 60Hz); 162L/min. (at 50Hz)
For Use With (Equipment) Vacuum Concentrator
Oil Capacity 0.75 L
Ultimate Vacuum Level 1.5 mtorr/1.95 x10³mbar
Vacuum Inlet Fitting 12.7 OD
Width (English) 6 in.
Width (Metric) 160 mm
Height (English) 11 in.
Height (Metric) 260 mm
Depth (English) 19 in.
Depth (Metric) 470 mm
Weight (English) 57 lb.
Weight (Metric) 26 kg
Electrical Requirements 230 V 50 Hz
Voltage 230 V
Frequency 50 Hz