Learn More
Borer Chemie™ deconex 26 PLUS
Product Code. 10319540
Change view
Click to view available options
Quantity:
1 x 7 kg
1 x 12 kg
1 x 35 kg
Unit Size:
12kg
35kg
7kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10319540 1 x 12 kg 12kg
10300261 1 x 7 kg 7kg
10451921 1 x 35 kg 35kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10319540 Supplier Borer Chemie™ Supplier No. 502300.00K10R
Cleaner and neutralizer for laboratory washers. Neutralizes carried-over alkaline residues, shortens rinsing phases and reduces water consumption. deconex 26 PLUS is also effective against acid-soluble residues.
  • Neutralizer and cleaner for automated use
  • Economical because highly concentrated, use 2 mL/L
  • Neutralizes carried-over alkaline residues, reduces rinsing phases
  • Reduces the consumption of rinsing water
  • Without surfactants, rinses off without leaving residues
  • Also as pre-rinse agent against acid-soluble deposits
  • Technical support by the manufacturer
Specifications
Form Liquid
pH ∼1.8 (1% Solution)
Composition Inorganic acid
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 12 kg
Volume (English) 2.2 gal.
Volume (Metric) 8.5 L
Weight (English) 26.5 lb.
Weight (Metric) 12 kg
Show More Show Less
compliance-icons
Product Identifier
  • deconex 26 PLUS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contain: phosphoric acid