Learn More
Borer Chemie™ deconex 26 PLUS
Product Code. 10300261
Change view
Click to view available options
Quantity:
1 x 7 kg
1 x 12 kg
1 x 35 kg
Unit Size:
12kg
35kg
7kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10300261 1 x 7 kg 7kg
10319540 1 x 12 kg 12kg
10451921 1 x 35 kg 35kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10300261 Supplier Borer Chemie™ Supplier No. 502300.00KK5R
Cleaner and neutralizer for laboratory washers. Neutralizes carried-over alkaline residues, shortens rinsing phases and reduces water consumption. deconex 26 PLUS is also effective against acid-soluble residues.
  • Neutralizer and cleaner for automated use
  • Economical because highly concentrated, use 2 mL/L
  • Neutralizes carried-over alkaline residues, reduces rinsing phases
  • Reduces the consumption of rinsing water
  • Without surfactants, rinses off without leaving residues
  • Also as pre-rinse agent against acid-soluble deposits
  • Technical support by the manufacturer
Specifications
Form Liquid
pH ∼1.8 (1% Solution)
Composition Inorganic acid
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 7 kg
Volume (English) 1.3 gal.
Volume (Metric) 5 L
Weight (English) 15.4 lb.
Weight (Metric) 7 kg
Show More Show Less
compliance-icons
Product Identifier
  • 7KG deconex 26 PLUS
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: phosphoric acid