Learn More
Borer Chemie™ deconex 22 PF
Product Code. 10732531
Click to view available options
Quantity:
6 x 1.5 kg
1 x 10 kg
Unit Size:
10kg
9kg
Product Code. 10732531
Brand: Borer Chemie™ 501500.00S10P
Universal powder cleaner for laboratory glassware and utensils. The ecological solution deconex 22 PF shows a very high cleaning performance without chlorine, no surfactants and no phosphates. An acid rinse with a deconex cleaner is recommended.
  • Powder cleaner for automated cleaning
  • Ecological, phosphate-free solution
  • Perfect results already with 3-5 g/L
  • Can be rinsed off without leaving residues
  • For consistent and flawless results
  • Wide cleaning spectrum
  • Without chlorine, EDTA, NTA and surfactants
  • Acid rinse with deconex cleaner recommended
  • Technical support by the manufacturer
Specifications
Form Powder
pH ∼12.1 (1% Solution)
Composition Alkalis, complexing agents, corrosion inhibitor
Concentration Concentrate
Container Type Bucket
Content And Storage 5°C to 25°C
Quantity 1 x 10 kg
Weight (English) 22 lb.
Weight (Metric) 10 kg
compliance-icons compliance-icons
Product Identifier
  • deconex 22 PF
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.