Learn More
Borer Chemie™ deconex 15 PF-x
Product Code. 10658619
Change view
Click to view available options
Quantity:
1 x 5 kg
1 x 10 kg
Unit Size:
10kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10658619 1 x 5 kg 5kg
10591938 1 x 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10658619 Supplier Borer Chemie™ Supplier No. 518000.00K05W
Liquid concentrate for manual cleaning of laboratory glassware and utensils in immersion baths, ultrasonic baths and pipette washers. deconex 15 PF-x is phosphate-free, ecological for versatile use, with high cleaning power and dispersing capacity.
  • Concentrate for manual cleaning
  • Also suitable for pipette washers
  • Ecological solution without phosphates
  • Without EDTA, NTA and chlorine
  • Highly productive, typical concentration: 10-20 mL/L
  • Rinses off without leaving residues
  • For consistent and error-free results
  • Extra wide cleaning spectrum
  • High swelling and dispersing capacity
  • Technical support by the manufacturer
Specifications
Form Liquid
pH ∼12.0 (1% Solution)
Composition Alkalis, complexing agents, surfactants, solubilizer
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 5 kg
Volume (English) 1.2 gal.
Volume (Metric) 4.5 L
Weight (English) 11 lb.
Weight (Metric) 5 kg
Show More Show Less
compliance-icons
Product Identifier
  • deconex 15 PF-x
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contain: potassium hydroxide