Learn More
Corning™ DeckWorks™ Tip Stations, Sterile
Product Code. 11921861 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
Unit Size:
Pack of 4800
Pack of 960
Product Code. Volume (Metric) unitSize
11921861 1 to 200 μL Pack of 960
11393665 1 to 200 μL Pack of 4800
11911861 0.1 to 10 μL Pack of 960
11383665 0.1 to 10 μL Pack of 4800
4 options
Product Code. 11921861
Brand: Corning™ 4144
Only null left
Add to basket
Add to basket
Designed for single and multichannel pipetting stations
Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip ­stations contain 960 tips, are available in extended-length 10 μL and traditional 200 μL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only. Applications include pre- and ­post-PCR procedures, cell culture, and assays requiring large tip quantities. Corning DeckWorks tip stations are offered both sterile and nonsterile.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1 to 200 μL
Sterility Sterile
Color Natural
Tip Style Round
Packaging Multi-pack
For Use With (Equipment) Universal Fit, Multichannel Pipetting Stations
Graduated Yes
Product Line DeckWorks