Learn More
Corning™ DeckWorks™ Tip Stations, Sterile
Product Code. 11393665 Shop All Corning Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
Unit Size:
Pack of 4800
Pack of 960
Product Code. 11393665
Brand: Corning™ 4144
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for single and multichannel pipetting stations
Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip ­stations contain 960 tips, are available in extended-length 10 μL and traditional 200 μL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only. Applications include pre- and ­post-PCR procedures, cell culture, and assays requiring large tip quantities. Corning DeckWorks tip stations are offered both sterile and nonsterile.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1 to 200 μL
Sterility Sterile
Color Natural
Tip Style Round
Packaging Multi-pack
For Use With (Equipment) Universal Fit, Multichannel Pipetting Stations
Graduated Yes
Product Line DeckWorks