Description
Corning DeckWorks tip station maximizes bench space while minimizing plastic waste. Tip stations contain 960 tips, are available in extended-length 10 μL and traditional 200 μL sizes, and are the ideal single and multi-channel pipetting stations. The 5-layer deck design (2 x 96) matches the microtiter plate format. This stable pipetting platform fits easily into sterile hood environments. Lids can be snap-positioned to expose a single side (96 tips) only. Applications include pre- and post-PCR procedures, cell culture, and assays requiring large tip quantities. Corning DeckWorks tip stations are offered both sterile and nonsterile.
Specifications
Specifications
| Volume (Metric) | 0.1 to 10 μL |
| Sterility | Sterile |
| Color | Natural |
| Tip Style | Round |
| Packaging | Multi-pack |
| For Use With (Equipment) | Universal Fit, Multichannel Pipetting Stations |
| Graduated | Yes |
| Product Line | DeckWorks |
| Material | Polypropylene |