Learn More
Corning™ DeckWorks Pipet Tips
Product Code. 11729325 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
1 to 300 μL
100 to 1000 μL
Color:
Natural
Yellow
Unit Size:
Pack of 3072
Pack of 3840
Pack of 768
Pack of 960
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color unitSize
11729325 1 to 200 μL Natural Pack of 960
11383645 1 to 300 μL Natural Pack of 3840
11749325 1 to 300 μL Natural Pack of 960
11363645 1 to 200 μL Natural Pack of 3840
11739325 1 to 200 μL Yellow Pack of 960
11373645 1 to 200 μL Yellow Pack of 3840
11719325 0.1 to 10 μL Natural Pack of 960
11353645 0.1 to 10 μL Natural Pack of 3840
11789315 100 to 1000 μL Natural Pack of 768
11393645 100 to 1000 μL Natural Pack of 3072
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11729325 Supplier Corning™ Supplier No. 4121
  • ONLINE EXCLUSIVE PRICE
    1045.00 NOK
    974.98 NOK / Pack of 960
    Save 70.02 NOK
    6% Off
View all promotions
In Stock
Add to basket
Add to basket
Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking
Corning universal fit tips in hinged DeckWorks racks are available in 10 μL, 200 μL, 300 μL, and 1,000 μL sizes. The 10 μL, 300 μL, and 1,000 μL tips are extended length and allow for improved access into microcentrifuge (1.5 mL) and standard conical bottom (15/50 mL) centrifuge tubes. Robust 96-tip racks are designed for repeated steam autoclave cycles and can be reused by removing the empty grey tip deck and replacing it with a new reload deck of 96 tips from the DeckWorks reload system. Corning racked 300 μL DeckWorks pipet tips are designed for all popular 300 μL multi-channel instruments.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Autoclavable
Certifications/Compliance Nonpyrogenic, Human DNA-free, RNase/DNase-free, Sterility Assurance Level 10-3
Clean Claims Nonpyrogenic, Human DNA-free, RNase/DNase-free, Sterility Assurance Level 10-3
For Use With (Application) Universal tip fits in hinged DeckWorks racks that allows for easy rack stacking
Format Hinged Rack
Tip Style Round
Packaging Hinged rack
Quantity 960/Pk., 3840/Cs.
Product Type Pipette Tips
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line DeckWorks