Learn More
Decanoic-10,10,10-d3 Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Decanoic-10,10,10-d3 Acid |
| CAS | 102611-15-4 |
| Molecular Formula | C10 D3 H17 O2 |
| Quantity | 10 mg |
| Synonym | 1-Decanoic Acid-d3, 1-Nonanecarboxylic Acid-d3, Capric Acid-d3, Caprinic Acid-d3, Caprynic Acid-d3, Decoic Acid-d3, Decylic Acid-d3, Emery 659-d3, Lunac 10-95-d3, Lunac 10-98-d3, Lunac 10-98E-d3, NAA 102-d3, NSC 5025-d3, Prifac 2906-d3, Prifac 296, -d3 n-Capric Acid-d3, n-Decanoic Acid-d3, n-Decoic Acid-d3, n-Decylic Acid-d3, Decanoic-10,10,10-d3 acid (9CI), 10,10,10-Trideuteriodecanoic acid, Decanoic acid-10,10,10-d3 |
| InChI Formula | InChI=1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)/i1D3 |
| SMILES | [2H]C([2H])([2H])CCCCCCCCC(=O)O |
| IUPAC Name | 10,10,10-trideuteriodecanoic acid |
| Molecular Weight (g/mol) | 175.28 |
| Formula Weight | 175.1652 |