missing translation for 'onlineSavingsMsg'
Learn More

Debio 0932, MedChemExpress

Product Code. 16435708
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16435708 10 mg 10mg
16468397 5 mg 5mg
16456196 100 mg 100mg
16451867 50 mg 50mg
16421086 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16435708 Supplier Medchem Express Supplier No. HY13469

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Debio 0932 (CUDC-305) is an orally active HSP90 inhibitor, with IC50s of 100 and 103 nM for HSP90α and HSP90β, respectively.

Debio 0932 (CUDC-305) is an orally active HSP90 inhibitor, with IC50s of 100 and 103 nM for HSP90α and HSP90β, respectively.

Specifications

Chemical Name or Material Debio 0932
CAS 1061318-81-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H30N6O2S
Quantity 10 mg
Synonym CUDC-305
Solubility Information DMSO : 75 mg/mL (169.46 mM; Need ultrasonic)
SMILES NC1=NC=CC2=C1N=C(SC3=C(N(C)C)C=C(OCO4)C4=C3)N2CCNCC(C)(C)C
Molecular Weight (g/mol) 442.58
Formula Weight 442.58
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.