missing translation for 'onlineSavingsMsg'
Learn More

DEAE-Dextran, chloride Form, MP Biomedicals™

Product Code. 11471002 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471002 100 g 100g
11451002 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471002 Supplier MP Biomedicals Supplier No. 0219513380

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 9015-73-0
MDL Number MFCD00081561
Synonym Diethylaminoethyl-dextrane
IUPAC Name Diethylaminoethyl dextran

Specifications

Physical Form Powder
Color White
Quantity 100 g
Formula Weight 500, 00
Chemical Name or Material DEAE-Dextran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.