Learn More
Trajan Scientific and Medical™ Deactivated (Phenyl) Fused Silica Tubing
Click to view available options
Overall Length (Metric):
10 m
5 m
Diameter (Metric) Inner:
0.25 mm
220 μm
Unit Size:
10m
5m
Description
- Chemically inert and thermally stable up to 380°C
- Suitable for organic and aqueous solvents
- Ideal for biotechnology applications
Specifications
Specifications
| For Use With | For Organic and Aqueous Solvents |
| Type | Tubing |
| Includes | 1/Pk. |
| Diameter (Metric) Inner | 220 μm |
| Diameter (Metric) Outer | 363 μm |
| Overall Length (Metric) | 10 m |
| Max. Operating Temperature (Metric) | Thermally stable up to 380°C |