Learn More
- Home
- Products
- Chromatography Supplies
- Chromatography Tubing
- Trajan Scientific and Medical™ Deactivated (Methyl) Fused Silica Tubing
Trajan Scientific and Medical™ Deactivated (Methyl) Fused Silica Tubing
Product Code. 15228803
Change view
Click to view available options
Overall Length (Metric):
10 m
2 m
25 m
5 m
Diameter (Metric) Inner:
0.05 mm
0.11 mm
0.125 mm
0.25 mm
25 μm
60 μm
75 μm
100 μm
110 μm
150 μm
170 μm
220 μm
250 μm
320 μm
530 μm
Diameter (Metric) Outer:
0.31 mm
0.363 mm
170 μm
220 μm
285 μm
300 μm
363 μm
430 μm
680 μm
Includes:
1/Pk.
5/Pk.
Unit Size:
10m
25m
2m
5m
Each
19 product options available for selection
| Product Code. | Overall Length (Metric) | Diameter (Metric) Inner | Diameter (Metric) Outer | Includes | unitSize |
|---|---|---|---|---|---|
| 15228803 | 2 m | 60 μm | 220 μm | 1/Pk. | 2m |
| 15258803 | 2 m | 0.05 mm | 0.363 mm | 1/Pk. | 2m |
| 12965751 | 5 m | 530 μm | 680 μm | 1/Pk. | 5m |
| 15298793 | 5 m | 250 μm | 363 μm | 1/Pk. | 5m |
| 15208813 | 10 m | 150 μm | 220 μm | 1/Pk. | 10m |
| 15288793 | 2 m | 25 μm | 285 μm | 1/Pk. | 2m |
| 15218803 | 2 m | 220 μm | 220 μm | 1/Pk. | 2m |
| 15278803 | 2 m | 110 μm | 170 μm | 1/Pk. | 2m |
| 15268813 | 5 m | 150 μm | 363 μm | 1/Pk. | 5m |
| 12766248 | 10 m | 220 μm | 363 μm | 1/Pk. | Each |
| 15258813 | 25 m | 220 μm | 363 μm | 1/Pk. | 25m |
| 12736248 | 2 m | 75 μm | 363 μm | 1/Pk. | Each |
| 12746248 | 25 m | 100 μm | 363 μm | 1/Pk. | Each |
| 12756248 | 10 m | 320 μm | 430 μm | 1/Pk. | Each |
| 15208803 | 10 m | 250 μm | 363 μm | 1/Pk. | 10m |
| 15208823 | 2 m | 170 μm | 300 μm | 5/Pk. | Each |
| 15248803 | 2 m | 0.125 mm | 0.363 mm | 1/Pk. | 2m |
| 15268803 | 2 m | 0.11 mm | 0.31 mm | 1/Pk. | 2m |
| 15268853 | 5 m | 0.25 mm | 0.363 mm | 1/Pk. | 25m |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Trajan methyl deactivated fused silica capillary tubing is an essential ingredient in maintaining a high performance chromatography system.
- Chemically inert and thermally stable up to 380°C
- Suitable for organic and aqueous solvents
- Ideal for biotechnology applications
Specifications
Specifications
| For Use With | Organic and Aqueous Solvents |
| Type | Tubing |
| Includes | 1/Pk. |
| Diameter (Metric) Inner | 60 μm |
| Diameter (Metric) Outer | 220 μm |
| Overall Length (Metric) | 2 m |
| Max. Operating Temperature (Metric) | Thermally stable up to 380°C |