Learn More
DDX4 Monoclonal Antibody (OTI1G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA813368S
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 17598224
  • 4065.15 NOK / 30µL
Estimated Shipment: 21-05-2024
Description
DDX4 Monoclonal antibody specifically detects DDX4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
DDX4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q61496, Q64060, Q9NQI0
Mouse
Protein A/G
RUO
13206, 310090, 54514
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI1G7
Unconjugated
DDX4
ATP-dependent RNA helicase DDX4; AV206478; Cvh; Ddx4; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (Asp-Glu-Ala-Asp) box polypeptide 4; DEAD box polypeptide 4; DEAD box protein 4; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4; DEAD-box helicase 4; MGC111074; Mvh; mvh / m'vasa; probable ATP-dependent RNA helicase DDX4; putative ATP-dependent RNA helicase DDX4; RVLG; VASA; Vasa homolog; vasa-like gene protein
Human recombinant protein fragment corresponding to amino acids 1-400 of human DDX4 (NP_077726) produced in E.coli.
30 μL
Primary
Human, Mouse, Rat
Antibody
IgG1