missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX19B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18285-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Spectrins are a family of filamentous cytoskeletal proteins that function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. Spectrins are composed of alpha and beta dimers that associate to form tetramers linked in a head-to-head arrangement. This gene encodes an alpha spectrin that is specifically expressed in nonerythrocytic cells. The encoded protein has been implicated in other cellular functions including DNA repair and cell cycle regulation. Mutations in this gene are the cause of early infantile epileptic encephalopathy-5. Alternate splicing results in multiple transcript variants.Specifications
DDX19B | |
Polyclonal | |
Unconjugated | |
DDX19B | |
DBP5, DDX19, DDX19B, DEAD box protein 19B, DEAD box RNA helicase DEAD5, RNAh | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11269, 234733, 690693 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UMR2 | |
Ddx19b | |
DDX19B Fusion Protein Ag13165 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |