missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDR2 Antibody (290804), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB25381W
This item is not returnable.
View return policy
Description
DDR2 Monoclonal antibody specifically detects DDR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-reported
Specifications
| DDR2 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 290804 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-reported | |
| CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKTMIG20a, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase | |
| Mouse myeloma cell line NS0-derived recombinant human DDR2, Gln24-Arg399, Accession # Q16832 | |
| 0.1 mL | |
| Cardiovascular Biology, CD Markers, Immunology, Neuroscience, Signal Transduction | |
| 4921 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction