Learn More
DDR1-IN-1, MedChemExpress
Product Code. 16456376
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16456376
Brand: Medchem Express HY13979
MedChemExpress DDR1-IN-1 is a potent and selective DDR1 receptor tyrosine kinase inhibitor with an IC50 of 105 nM; 4-fold less potent for DDR2 (IC50 = 413 nM).
DDR1-IN-1 is a potent and selective DDR1 receptor tyrosine kinase inhibitor with an IC50 of 105 nM
Specifications
Chemical Name or Material DDR1-IN-1
CAS 1449685-96-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H31F3N4O3
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (180.97 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C)C(OC2=CC3=C(NC(C3)=O)C=C2)=C1)C4=CC=C(CN5CCN(CC)CC5)C(C(F)(F)F)=C4
Molecular Weight (g/mol) 552.59
Formula Weight 552.59
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.