Learn More
Abnova™ DDIT3 (Human) Recombinant Protein
Description
- Sequence: MAAESLPFSFGTLSSWELEAWYEDLQEVLSSDENGGTYVSPPGNEEEESKIFTTLDPASLAWLTEEEPEPAEVTSTSQSPHSPDSSQSSLAQEEEEEDQGRTRKRKQSGHSPARAGKQRMKEKEQENERKVAQLAEENERLKQEIERLTREVEATRRALIDRMVNLHQA
Specifications
Specifications
| Accession Number | AAH03637.1 |
| Gene ID (Entrez) | 1649 |
| Name | DNA-damage-inducible transcript 3 |
| Preparation Method | Wheat germ expression system |
| Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
| Quantity | 25 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | CEBPZ, CHOP, CHOP10, GADD153, MGC4154 |
| Gene Symbol | DDIT3 |
| Species | Wheat Germ (in vitro) |
| Show More |