missing translation for 'onlineSavingsMsg'
Learn More

DDI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13968-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16892414

  • 1281.92 NOK / 20µL
Estimated Shipment: 20-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ddi1 (DNA-damage inducible 1)appears to release substrate from an ubiquitination complex, making the substrate available for deubiquitination to facilitate proteasomal degradation. Ddi1 could be a potential selective target for lung cancer treatment. Ddi1 runs as a doublet of 54/56 kDa on acrylamide gels besides the calculated MW 44 kDa.
Specifications

Specifications

DDI1
Polyclonal
Unconjugated
DDI1
DDI1, Protein DDI1 homolog 1
Rabbit
Antigen Affinity Chromatography
RUO
367012, 414301, 71829
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A0JPP7, Q8WTU0, Q9DAF3
Ddi1
DDI1 Fusion Protein Ag4992
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers