missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDHD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25203-1-AP-20UL
This item is not returnable.
View return policy
Description
DDHD2, also known as KIAA0725p, is a member of the intracellular phospholipase A1 (PLA1) protein family which comprises a group of enzymes that hydrolyze the sn-1 ester bond of phospholipids, producing 2-acyl-lysophospholipids and fatty acids. DDHD2 prefers phosphatidic acid as substrate and has a role in efficient membrane trafficking from the Golgi apparatus to the plasma membrane. The gene of DDHD2 maps to chromosome 8p11.23, and encodes a 711-amino-acid protein with a calculated molecular mass of 81 kDa. It has been reported that DDHD2 immunoblot analysis of various tissue extracts revealed two bands of 90 and 85 kDa.Specifications
| DDHD2 | |
| Polyclonal | |
| Unconjugated | |
| DDHD2 | |
| DDHD domain containing 2, DDHD2, KIAA0725, KIAA0725p, Phospholipase DDHD2, SAMWD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23259, 680971, 72108 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O94830, Q80Y98 | |
| Ddhd2 | |
| DDHD2 Fusion Protein Ag18373 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction