missing translation for 'onlineSavingsMsg'
Learn More

DCUN1D3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1710.33 NOK - 4836.10 NOK

Specifications

Antigen DCUN1D3
Concentration 0.33 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16804855
View Documents
Proteintech
26356-1-AP-20UL
20 μL
1710.33 NOK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16894845
View Documents
Proteintech
26356-1-AP-150UL
150 μL
4836.10 NOK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DCUN1D3 contains 1 DCUN1 domain. The exact function of DCUN1D3 remains unknown.
Specifications

Specifications

DCUN1D3
Western Blot
Unconjugated
Rabbit
Human
Q8IWE4
123879
DCUN1D3 Fusion Protein Ag23730
Primary
-20°C
DCUN1D3
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
44M2.4, DCN1 like protein 3, DCUN1D3
DCUN1D3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.