Learn More
DBR1 Mouse anti-Human, Unconjugated, Clone: 3B5A7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66951-1-IG-20UL
 View more versions of this product
Product Code. 16809275
  • 1710.33 NOK / 20µL
Description
The DBR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DBR1. This antibody recognizes human, mouse, rat, pig antigen. The DBR1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
DBR1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
3B5A7
Unconjugated
20 μL
51163
IgG2b