Learn More
DBR1 Mouse anti-Human, Unconjugated, Clone: 3B5A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66951-1-IG-20UL
Description
The DBR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DBR1. This antibody recognizes human, mouse, rat, pig antigen. The DBR1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| DBR1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 3B5A7 | |
| Unconjugated | |
| 20 μL | |
| 51163 | |
| IgG2b |