missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBI Monoclonal Antibody (OTI3A10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813117
This item is not returnable.
View return policy
Description
DBI Monoclonal antibody specifically detects DBI in Human samples. It is validated for Western BlotSpecifications
| DBI | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07108 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1622 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3A10 | |
| Unconjugated | |
| DBI | |
| ACBD 1; ACBD1; ACBP; Acoabp3; acyl coenzyme A binding protein; acyl-CoA binding protein; acyl-CoA-binding protein; acyl-Coenzyme A binding domain containing 1; CCK RP; CCKRP; CCK-RP; cholecystokinin-releasing peptide, trypsin-sensitive; Dbi; diazepam binding inhibitor; Diazepam binding inhibitor (GABA receptor modulator acyl-Coenxyme A binding protein); diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein); Diazepam binding inhibitor (GABA receptor modulator, acyl-Coenxyme A binding protein); diazepam binding inhibitor (GABA receptor modulator, acyl-Coenzyme A binding protein); diazepam binding inhibitor, acyl-CoA binding protein; diazepam binding inhibitor, splice form 1b; diazepam-binding inhibitor; Endozepine; EP; GABA receptor modulator; LRRGT00046; MGC70414; octadecaneuropeptide; ODN; RNACOABP3; Triakontatetraneuropeptide; Ttn; valium | |
| Human recombinant protein fragment corresponding to amino acids 2-104 of human DBI (NP_065438) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction