Learn More
Emma Safety Footwear Daytona S1P Sandal
Daytona is one of Emma's most breathable shoes. It's an ideal choice for warm, enclosed environments such as those in the Automotive industry.
Brand: Emma Safety Footwear MM78754744
Additional Details : Weight : 1.11000kg
Description
- Metal Free
- Breathable and incredibly comfortable
- It's not hard to see why the Daytona is one of Emma's most breathable shoes. It's an ideal choice for warm, enclosed environments such as those in the Automotive industry.
- It features Emma's unique Hydro-Tec™ antibacterial technology, removing moisture and perspiration instantly.
- Synthetic, reinforced toes and super swift velcro fasteners make the Daytona an ideal choice for all active occupations but especially for people working in Warehousing and Logistics.
- Industries
- Logistics & transport
- Electronics
- Manufacturing
- Safety shoe
- S1P-Safetyshoe with sole plate for dry work
Specifications
ISO 20345:2011 | |
Black, Gray | |
Hydro-Tec Antibacterial | |
44 | |
Safety Shoe |
Velcro | |
Logistics and Transport, Electronics, Manufacturing | |
Nubuck | |
Standard width |