missing translation for 'onlineSavingsMsg'
Learn More

Daunomycin, 95%, MP Biomedicals™

Product Code. 11452172 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452172 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11452172 Supplier MP Biomedicals Supplier No. 0215077710
  • ONLINE EXCLUSIVE PRICE
    2590.00 NOK
    2464.64 NOK / 10mg
    Save 125.36 NOK
    4% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sequence-specific double-stranded DNA intercalating agent used in antileukemic, photostability, and drug metabolism research.

Specifications

Type Daunomycin
Physical Form Powder
CAS 23541-50-6
Color Red
Quantity 10 mg
Assay Percent Range 0.95
Molecular Formula C27H30ClNO10
Synonym Daunorubicin, Leukaemomycin C
InChI Key GUGHGUXZJWAIAS-QQYBVWGSSA-N
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl
IUPAC Name (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
Molecular Weight (g/mol) 563.984
PubChem CID 62770
Formula Weight 564
Grade Not Available
Chemical Name or Material Daunomycin, For HPLC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.