Learn More
Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers
Product Code. 11913947 Shop All Thermo Scientific Products
Click to view available options
Type:
RS-232 Output
RS-485 Output
Voltage:
0/1 V
0/5 V
4-20mA
Unit Size:
Each
Product Code. 11913947
Brand: Thermo Scientific™ 195898
Only null left
Add to basket
Add to basket
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
RS-232:
  • RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
  • RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
TRUSTED_SUSTAINABILITY
Specifications
Type RS-485 Output
For Use With (Equipment) Revco™ CxF, HERAfreeze™ HFC, Model 1535 Monitor/Alarm System, TSC Series
Monitoring Options Alarm, Recorder
Voltage 0/1 V