Learn More
Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers
Shop All Thermo Scientific ProductsDescription
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
RS-232:
- RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
- RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
Specifications
Specifications
| Type | RS-485 Output |
| For Use With (Equipment) | Revco™ CxF, HERAfreeze™ HFC, Model 1535 Monitor/Alarm System, TSC Series |
| Monitoring Options | Alarm, Recorder |
| Voltage | 0/5 V |