Learn More
SI Analytics™ Data Cable
Product Code. 10161971 Shop All SI Analytics Products
Change view
Click to view available options
Length (Metric):
1.5 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10161971 1.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10161971 Supplier SI Analytics™ Supplier No. 285223578
  • ONLINE EXCLUSIVE PRICE
    907.00 NOK
    849.27 NOK / Pack of 1
    Save 57.73 NOK
    6% Off
 Restricted Item
In Stock
Data cable, 1.5 m length for use with Titronic™ and TitroLine™ titrators
  • Length 1.5 m
  • For use with Titronic 300, Titronic 500, TitroLine 5000, TitroLine 6000, 7000, 7750, 7800, 7500 KF, 7500 KF trace titrators
TRUSTED_SUSTAINABILITY
Specifications
Description Data cable 1.5 m
Length (Metric) 1.5 m