Learn More
Fisherbrand™ Traceable™ Data Acquisition Software
Product Code. p-7217108 Shop All Cole-Parmer Products
Click to view available options
Includes:
Cailbration, Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
Unit Size:
Each
Product Code. 17156324
Brand: Fisherbrand™ 9008021
Only null left
Add to basket
Add to basket
Complete Data Acquisition Software enables measurement readings to be displayed and stored on a PC from up to 8 devices at a time.
  • Display and store measurement readings on a PC from up to 8 devices at a time
  • Data capture runs in the background, enabling users to work on other programs simultaneously
  • Set audible and visual notification alarms, or immediately send an email if an over/under condition occurs
  • Software can be used to connect a meter directly to PC via cable
  • Check software specification tables for meter compatibility
Specifications
Type Data Acquisition Software
Display Type No Display
Description Data Acquisition Software with USB Cable and Calibration
Includes Cailbration, Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
For Use With (Equipment) 90080-09, 03313-65, 03313-66, 37804-06, 37804-07 (Traceable™ 4000, 4080, 4085, 6412, 6413)