missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DARS (Human) Recombinant Protein
Human DARS full-length ORF ( AAH00629, 1 a.a. - 501 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001615-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPSASASRKSQEKPREIMDAAEDYAKERYGISSMIQSQEKPDRVLVRVRDLTIQKADEVVWVRARVHTSRAKGKQCFLVLRQQQFNVQALVAVGDHASKQMVKFAANINKESIVDVEGVVRKVNQKIGSCTQQDVELHVQKIYVISLAEPRLPLQLDDAVRPEAEGEEEGRATVNQDTRLDNRVIDLRTSTSQAVFRLQSGICHLFRETLINKGFVEIQTPKIISAASEGGANVFTVSYFKNNAYLAQSPQLYKQMCICADFEKVFSIGPVFRAEDSNTHRHLTEFVGLDIEMAFNYHYHEVMEEIADTMVQIFKGLQERFQTEIQTVNKQFPCEPFKFLEPTLRLEYCEALAMLREAGVEMGDEDDLSTPNEKLLGHLVKEKYDTDFYILDKYPLAVRPFYTMPDPRNPKQSNSYDMFMRGEEILSGAQRIHDPQLLTERALHHGIDLEKIKAYIDSFRFGAPPHAGGGIGLERVTMLFLGLHNVRQTSMFPRDPKRLTP
Specifications
AAH00629 | |
aspartyl-tRNA synthetase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DARS | |
GST |
1615 | |
Wheat germ expression system | |
25 μg | |
DKFZp781B11202, MGC111579 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |