missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DAPK3 (Human) Recombinant Protein
Human DAPK3 full-length ORF ( NP_001339.1, 1 a.a. - 454 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001613-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSTFRQEDVEDHYEMGEELGSGQFAIVRKCRQKGTGKEYAAKFIKKRRLSSSRRGVSREEIEREVNILREIRHPNIITLHDIFENKTDVVLILELVSGGELFDFLAEKESLTEDEATQFLKQILDGVHYLHSKRIAHFDLKPENIMLLDKNVPNPRIKLIDFGIAHKIEAGNEFKNIFGTPEFVAPEIVNYEPLGLEADMWSIGVITYILLSGASPFLGETKQETLTNISAVNYDFDEEYFSNTSELAKDFIRRLLVKDPKRRMTIAQSLEHSWIKAIRRRNVRGEDSGRKPERRRLKTTRLKEYTIKSHSSLPPNNSYADFERFSKVLEEAAAAEEGLRELQRSRRLCHEDVEALAAIYEEKEAWYREESDSLGQDLRRLRQELLKTEALKRQAQEEAKGALLGTSGLKRRFSRLENRYEALAKQVASEMRFVQDLVRALEQEKLQGVECGLR
Specifications
NP_001339.1 | |
death-associated protein kinase 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAPK3 | |
GST |
1613 | |
Wheat germ expression system | |
25 μg | |
FLJ36473, ZIP, ZIPK | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |