missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DAP (Human) Recombinant Protein
Human DAP full-length ORF ( AAH02726, 1 a.a. - 102 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001611-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSSPPEGKLETKAGHPPAVKAGGMRIVQKHPHTGDTKEEKDKDDQEWESPSPPKPTVFISGVIARGDKDFPPAAAQVAHQKPHASMDKHPSPRTQHIQQPRK
Specifications
AAH02726 | |
death-associated protein | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAP | |
GST |
1611 | |
Wheat germ expression system | |
10 μg | |
MGC99796 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |