Learn More
Danirixin, MedChemExpress
Product Code. 16497706
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16497706
Brand: Medchem Express HY19768
MedChemExpress Danirixin is a selective, and reversible CXCR2 antagonist, with IC50 of 12.5 nM for CXCL8.
Danirixin is a selective, and reversible CXCR2 antagonist, with IC50 of 12.5 nM for CXCL8.
Specifications
Chemical Name or Material Danirixin
CAS 954126-98-8
Color White
Purity Grade Notes Research
Molecular Formula C19H21ClFN3O4S
Quantity 100 mg
Synonym GSK1325756
Solubility Information DMSO : 8 mg/mL (18.10 mM; Need ultrasonic and warming)
SMILES O=C(NC1=CC=C(Cl)C(S([C@H]2CCCNC2)(=O)=O)=C1O)NC3=C(C)C(F)=CC=C3
Molecular Weight (g/mol) 441.9
Formula Weight 441.9
Percent Purity 98.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.