missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ DANI Liners

Provides better analysis for a wide range of compounds. Trajan Scientific and Medical DANI Liners has unique deactivation process for SGE standard inlets.

Brand:  Trajan Scientific and Medical™ 092501

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15219043

  • 1725.00 NOK / Pack of 5
Estimated Shipment: 08-07-2024
to see stock.
Add to basket

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with DANI instruments
Specifications

Specifications

Straight-through Liner
DANI Liners
70 mm
6 mm
Deactivated Liners For DANI GCs
5/Pk.
4 mm
Straight Liner for DANI
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers