missing translation for 'onlineSavingsMsg'
Learn More

Danegaptide Hydrochloride, MedChemExpress

Product Code. p-200098263
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16457007

Brand: Medchem Express HY10913A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Danegaptide Hydrochloride (GAP-134 Hydrochloride) is a potent, selective and orally active gap-junction modifier with an antiarrhythmic effect.

Danegaptide Hydrochloride (GAP-134 Hydrochloride) is a potent, selective and orally active gap-junction modifier with an antiarrhythmic effect.

Specifications

Chemical Name or Material Danegaptide Hydrochloride
CAS 943133-81-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H18ClN3O4
Quantity 5 mg
Synonym GAP-134 Hydrochloride ZP 1609 Hydrochloride
Solubility Information DMSO : ≥ 55 mg/mL (167.81 mM) ∣H2O : ≥ 50 mg/mL (152.55 mM)
SMILES O=C(N[C@H]1CN(C(CN)=O)[C@H](C(O)=O)C1)C2=CC=CC=C2.[H]Cl
Molecular Weight (g/mol) 327.76
Formula Weight 327.76
Percent Purity 97.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.