Learn More
Danegaptide Hydrochloride, MedChemExpress
Product Code. 16457007
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16457007 5 mg 5mg
16440617 50 mg 50mg
16434288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16457007 Supplier Medchem Express Supplier No. HY10913A
MedChemExpress Danegaptide Hydrochloride (GAP-134 Hydrochloride) is a potent, selective and orally active gap-junction modifier with an antiarrhythmic effect.
Danegaptide Hydrochloride (GAP-134 Hydrochloride) is a potent, selective and orally active gap-junction modifier with an antiarrhythmic effect.
Specifications
Chemical Name or Material Danegaptide Hydrochloride
CAS 943133-81-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H18ClN3O4
Quantity 5 mg
Synonym GAP-134 Hydrochloride ZP 1609 Hydrochloride
Solubility Information DMSO : ≥ 55 mg/mL (167.81 mM) ∣H2O : ≥ 50 mg/mL (152.55 mM)
SMILES O=C(N[C@H]1CN(C(CN)=O)[C@H](C(O)=O)C1)C2=CC=CC=C2.[H]Cl
Molecular Weight (g/mol) 327.76
Formula Weight 327.76
Percent Purity 97.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation