Learn More
Daminozide, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Daminozide |
| CAS | 1596-84-5 |
| Molecular Formula | C6 H12 N2 O3 |
| Quantity | 25 g |
| Synonym | Butanedioic acid, mono(2,2-dimethylhydrazide) (9CI), Succinic acid, mono(2,2-dimethylhydrazide) (8CI), Alar, Alar 85, Aminozid, Aminozide, B 995, B-Nine, Bijiu, DIMG, DMASA, DYaK, Daminozide, Dazide, Dimas, Kylar, N-(Dimethylamino)succinamic acid, SADH, Succinic N',N'-dimethylhydrazide, Succinic acid 2,2-dimethylhydrazide, Succinic acid N,N-dimethylhydrazide |
| InChI Formula | InChI=1S/C6H12N2O3/c1-8(2)7-5(9)3-4-6(10)11/h3-4H2,1-2H3,(H,7,9)(H,10,11) |
| SMILES | CN(C)NC(=O)CCC(=O)O |
| IUPAC Name | 4-(2,2-dimethylhydrazinyl)-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 160.17 |
| Formula Weight | 160.0848 |