Learn More
Daminozide, MedChemExpress
Product Code. 16498457
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16498457
Brand: Medchem Express HY13643
MedChemExpress Daminozide, a plant growth regulator, is a selective inhibitor of the human KDM2/7 histone demethylases, with IC50s of 0.55, 1.5 and 2.1 μM for PHF8, KDM2A, and KIAA1718, respectively. Daminozide has >100-fold selectivity for KDM2/7 subfamily versus other demethylase subfamily members tested.
Daminozide, a plant growth regulator, is a selective inhibitor of the human KDM2/7 histone demethylases, with IC50s of 0.55, 1.5 and 2.1 μM for PHF8, KDM2A, and KIAA1718, respectively. Daminozide has >100-fold selectivity for KDM2/7 subfamily versus other demethylase subfamily members tested.
Specifications
Chemical Name or Material Daminozide
Physical Form Solid
CAS 1596-84-5
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C6H12N2O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 106.67 mg/mL (665.98 mM; Need warming) ∣H2O : 100 mg/mL (624.34 mM; Need ultrasonic)
SMILES O=C(NN(C)C)CCC(O)=O
Molecular Weight (g/mol) 160.17
Formula Weight 160.17
Percent Purity 95.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.