missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12224-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the family of dihydroxyacetone kinases, which have a protein structure distinct from other kinases. The product of this gene phosphorylates dihydroxyacetone, and also catalyzes the formation of riboflavin 4′,5′-phosphate (aka cyclin FMN) from FAD. Several alternatively spliced transcript variants have been identified, but the full-length nature of only one has been determined.Specifications
DAK | |
Polyclonal | |
Unconjugated | |
TKFC | |
DAK, DHA kinase, FMN cyclase, NET45 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
225913, 26007, 361730 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3LXA3, Q4KLZ6, Q8VC30 | |
Tkfc | |
DAK Fusion Protein Ag2861 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |