missing translation for 'onlineSavingsMsg'
Learn More

Dabigatran etexilate, MedChemExpress

Product Code. p-200098424
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16496477 5 mg 5mg
16494816 100 mg 100mg
16470137 50 mg 50mg
16433758 10 mg 10mg
4 options
This item is not returnable. View return policy

Product Code. 16496477

Brand: Medchem Express HY102745mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dabigatran etexilate (BIBR 1048) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.

Dabigatran etexilate (BIBR 1048) is an orally active prodrug of Dabigatran (a direct inhibitor of thrombin). Dabigatran etexilate has anticoagulant effects and is used for the prophylaxis of venousthromboembolism and stroke due to atrial fibrillation.

Specifications

Chemical Name or Material Dabigatran etexilate
CAS 211915-06-9
Color White
Purity Grade Notes Research
Molecular Formula C34H41N7O5
Quantity 5 mg
Synonym BIBR 1048
Solubility Information DMSO : ≥ 100 mg/mL (159.30 mM)
SMILES O=C(N(C1=NC=CC=C1)CCC(OCC)=O)C2=CC=C3C(N=C(N3C)CNC4=CC=C(C(NC(OCCCCCC)=O)=N)C=C4)=C2
Molecular Weight (g/mol) 627.73
Formula Weight 627.73
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.