Learn More
3M™ Secure Click™ Filter with Dual Flow
3M™ Secure Click™ Cartridges are CE approved against certain organic vapours with a boiling point above 65 °C.
Brand: 3M™ 7100153231
631.00 NOK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.21000kg
Description
Use our cartridge with 3M™ Secure Click™ Reusable Respirators 800 Series. New dual flow cartridges and filters allow for easier breathing.Specifications
CE Approved | |
A2 Cartridge | |
D8055 |
Secure Click™ | |
Cartridge | |
64/Cs. |