missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ D500 Foam Cut Protection Gloves

Catalog Number 17290471
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Bamboo TwinFlex™ technology provides level D cut protection, while SoftGrip foam coating on the palm and fingertips enhances grip capabilities

  • Bamboo TwinFlex™ Technology provides level D cut protection
  • Highly abrasion resistant for extended glove life and cost efficiency
  • High-performance elastomer (HPE) and SoftGrip foam coating for enhanced grip
  • Increased tactile sensitivity for part handling & assembly
  • 3D Ergo Technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material HPE
Color Green, Black
Material Dyneema™, Polyamide, Bamboo
Size 7
EN388 Standard 4X42D
For Use With (Application) Automotive, Construction, Glass Handling, Maintenance, and Touchscreens
Gender Unisex
Glove Cuff Style Knit
Series D500 Foam
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'