missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ D-Val-Leu-Lys-7-amino-4-methylcoumarin

Product Code. p-7139392 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10mg
25mg
Unit Size:
10mg
25mg
This item is not returnable. View return policy

Product Code. 11494686

Brand: MP Biomedicals™ 03AMC08110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-Val-Leu-Lys-7-amino-4-methylcoumarin

  • It is used for assay of tissue plasminogen activator.
  • The AMC fluorogenic substrates are widely used for routine assays of peptidases.
  • A short peptide sequence is conjugated to 7-AMINO-4-METHYLCOUMARIN, which is released upon cleavage by peptidase.
  • The release of 7-AMINO-4-METHYLCOUMARIN is then measured spectrophotometrically.
  • The D-VAL-LEU-LYS-7-AMINO-4-METHYLCOUMARIN is used as s substrate for Plasmin
TRUSTED_SUSTAINABILITY

Specifications

Color White
Form Solid
For Use With (Application) Apoptosis, Cell Viability Analysis, Cell Signalling
Quantity 10mg
Molecular Weight (g/mol) 515.4 (Not including 2TFA salt)
Product Type Proteins, Enzymes & Peptides
Storage Requirements Store at Room Temperature (15-30°C). Keep cool and dry.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.