Learn More
Thermo Scientific Chemicals D-(+)-Trehalose dihydrate, 99%
Product Code. p-3173071
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11404027
Brand: Thermo Scientific Chemicals A19434.06
Request Bulk or Custom Format
Edge
CAS: 6138-23-4 | C12H22O11 | 342.30 g/mol
D-(+)-Trehalose dihydrate is used as a food additive in many processed foods and pharmaceutical excipients. It is widely used as a direct replacement of conventional sweeteners and protects the cells from various environmental stresses. It finds application in the treatment of Huntington's disease and Parkinson's disease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water and hot alcohol.
Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6138-23-4
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.297
MDL Number MFCD00071594
InChI Key HDTRYLNUVZCQOY-LIZSDCNHSA-N
Synonym trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose
PubChem CID 7427
ChEBI CHEBI:16551
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(C(C(C(O2)CO)O)O)O)O)O)O)O
Specifications
Melting Point 97°C to 99°C
Assay Percent Range 0.99
Quantity 5 g
Beilstein 5322018
Sensitivity Hygroscopic
Solubility Information Soluble in water and hot alcohol.
Formula Weight 378.33 (342.29 Anhydrous)
Chemical Name or Material D-(+)-Trehalose dihydrate