Learn More
D-Serine Benzyl Ester Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | D-Serine Benzyl Ester Hydrochloride |
| CAS | 151651-44-4 |
| Molecular Formula | C10H13NO3 . HCl |
| Quantity | 10 g |
| Synonym | D-Serine','phenylmethyl ester','hydrochloride (1:1) (ACI), D-Serine','phenylmethyl ester','hydrochloride (9CI), D-Serine benzyl ester hydrochloride, Benzyl (R)-(+)-2-amino-3-hydroxypropanoate hydrochloride, (R)-Benzyl 2-amino-3-hydroxypropanoate hydrochloride, Benzyl D-serinate hydrochloride, Benzyl (2R)-2-amino-3-hydroxypropanoate hydrochloride, D-Serine phenylmethyl ester hydrochloride |
| InChI Formula | InChI=1S/C10H13NO3.ClH/c11-9(6-12)10(13)14-7-8-4-2-1-3-5-8;/h1-5,9,12H,6-7,11H2;1H/t9-;/m1./s1 |
| SMILES | Cl.N[C@H](CO)C(=O)OCc1ccccc1 |
| IUPAC Name | benzyl (2R)-2-amino-3-hydroxypropanoate;hydrochloride |
| Molecular Weight (g/mol) | 231.68 |
| Formula Weight | 231.0662 |